ENA181912121 Enamine

[3-methoxy-4-(2,2,2-trifluoroethoxy)phenyl]methanamine

  • MDL number MFCD09049307
  • Cas Number : 926224-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-36821

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36821
Availability details for
Quantity
  • Availability
  • Add to Cart