ENA181912127 Enamine

4-(3-hydroxypropyl)-1-phenyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD11099432
  • Cas Number : 1152610-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36829

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36829
Availability details for
Quantity
  • Availability
  • Add to Cart