ENA181912139 Enamine

2-(cyclopentylformamido)acetic acid

  • MDL number MFCD08445065
  • Cas Number : 82291-54-1
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36845

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36845
Availability details for
Quantity
  • Availability
  • Add to Cart