ENA181912147 Enamine

2-[(1H-pyrazol-1-yl)methyl]aniline

  • MDL number MFCD08572198
  • Cas Number : 956533-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36853

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36853
Availability details for
Quantity
  • Availability
  • Add to Cart