ENA183041994 Enamine

4-[(3-chloropropanoyl)amino]-N-(2-methoxyphenyl)benzamide

  • MDL number MFCD11505355
  • Cas Number : 1087791-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 193...195
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25841

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25841
Availability details for
Quantity
  • Availability
  • Add to Cart