ENA183041995 Enamine

2-chloro-N-methoxyacetamide

  • MDL number MFCD11505358
  • Cas Number : 36851-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26804

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26804
Availability details for
Quantity
  • Availability
  • Add to Cart