ENA183042010 Enamine

2-(5-methylfuran-2-yl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD11505390
  • Cas Number : 1087792-53-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39221

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39221
Availability details for
Quantity
  • Availability
  • Add to Cart