ENA183042012 Enamine

1-(3-nitrophenyl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD11505396
  • Cas Number : 1152836-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39238

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39238
Availability details for
Quantity
  • Availability
  • Add to Cart