ENA183042019 Enamine

N-[3-(1-aminoethyl)phenyl]methanesulfonamide hydrochloride

  • MDL number MFCD11505404
  • Cas Number : 1170616-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39285
Availability details for
Quantity
  • Availability
  • Add to Cart