ENA183042023 Enamine

6-chloro-8-methylquinolin-5-amine

  • MDL number MFCD11505409
  • Cas Number : 50358-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39310

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39310
Availability details for
Quantity
  • Availability
  • Add to Cart