ENA183042036 Enamine

4-{3,4,6-trimethyl-1H-pyrazolo[3,4-b]pyridin-1-yl}aniline

  • MDL number MFCD11505430
  • Cas Number : 1087792-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39404

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39404
Availability details for
Quantity
  • Availability
  • Add to Cart