ENA183042038 Enamine

1-benzyl-5-(thiophen-2-yl)imidazolidine-2,4-dione

  • MDL number MFCD11505434
  • Cas Number : 875248-15-0
  • Molecular Weight : 272.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-39412

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39412
Availability details for
Quantity
  • Availability
  • Add to Cart