ENA183042040 Enamine

(3-ethoxypropyl)(2,2,2-trifluoroethyl)amine hydrochloride

  • MDL number MFCD11505440
  • Cas Number : 1171876-76-8
  • Molecular Weight : 221.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39429

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39429
Availability details for
Quantity
  • Availability
  • Add to Cart