ENA183042043 Enamine

N-phenyl-1,4-diazepane-1-carboxamide hydrochloride

  • MDL number MFCD11505444
  • Cas Number : 1171206-49-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-39450

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39450
Availability details for
Quantity
  • Availability
  • Add to Cart