ENA183042050 Enamine

5-amino-N-(4-bromophenyl)-2-fluorobenzenesulfonamide

  • MDL number MFCD11505452
  • Cas Number : 1087784-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161...163
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39477

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39477
Availability details for
Quantity
  • Availability
  • Add to Cart