ENA183042053 Enamine

2-amino-2-(3,4,5-trimethoxyphenyl)acetonitrile hydrochloride

  • MDL number MFCD11505455
  • Cas Number : 348143-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39495

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39495
Availability details for
Quantity
  • Availability
  • Add to Cart