ENA183042054 Enamine

(1E)-1-(3-methyl-1-benzofuran-2-yl)ethanone oxime

  • MDL number MFCD11505456
  • Cas Number : 344287-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39502

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39502
Availability details for
Quantity
  • Availability
  • Add to Cart