ENA183042055 Enamine

2-amino-N-(1,1-dioxo-1lambda6-thiolan-3-yl)acetamide hydrochloride

  • MDL number MFCD11505457
  • Cas Number : 1171437-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-39506

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39506
Availability details for
Quantity
  • Availability
  • Add to Cart