ENA183042057 Enamine

3-[1-(ethylamino)ethyl]phenol hydrobromide

  • MDL number MFCD11505460
  • Cas Number : 1170975-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39510

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39510
Availability details for
Quantity
  • Availability
  • Add to Cart