ENA183042058 Enamine

ethyl[1-(3-methoxyphenyl)ethyl]amine hydrochloride

  • MDL number MFCD11505461
  • Cas Number : 1170204-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39517

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39517
Availability details for
Quantity
  • Availability
  • Add to Cart