ENA183042071 Enamine

3-amino-N-(1,1-dioxo-1lambda6-thiolan-3-yl)propanamide hydrochloride

  • MDL number MFCD11505476
  • Cas Number : 1172392-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39564

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39564
Availability details for
Quantity
  • Availability
  • Add to Cart