ENA183042077 Enamine

3-[(pyridin-2-yl)methanesulfonyl]propanoic acid hydrochloride

  • MDL number MFCD11505484
  • Cas Number : 1171061-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39589

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39589
Availability details for
Quantity
  • Availability
  • Add to Cart