ENA183042081 Enamine

[(naphthalen-1-yl)methyl]hydrazine dihydrochloride

  • MDL number MFCD11505488
  • Cas Number : 1177306-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39601

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39601
Availability details for
Quantity
  • Availability
  • Add to Cart