ENA183042085 Enamine

2-amino-N-[(pyridin-2-yl)methyl]acetamide dihydrochloride

  • MDL number MFCD11505495
  • Cas Number : 1170069-66-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39654

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39654
Availability details for
Quantity
  • Availability
  • Add to Cart