ENA183042090 Enamine

1-(5-nitrothiophene-2-carbonyl)piperazine hydrochloride

  • MDL number MFCD11505503
  • Cas Number : 1171194-70-9
  • Molecular Weight : 277.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-39688

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39688
Availability details for
Quantity
  • Availability
  • Add to Cart