ENA183042092 Enamine

({2,6-dimethylimidazo[2,1-b][1,3]thiazol-5-yl}methyl)(methyl)amine dihydrochloride

  • MDL number MFCD11505506
  • Cas Number : 1177362-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39706

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39706
Availability details for
Quantity
  • Availability
  • Add to Cart