ENA183042097 Enamine

N-(3-aminopropyl)-N-[(pyridin-3-yl)methyl]methanesulfonamide dihydrochloride

  • MDL number MFCD11505511
  • Cas Number : 1172233-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39727

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39727
Availability details for
Quantity
  • Availability
  • Add to Cart