ENA183042103 Enamine

2-(hydrazinomethyl)imidazo[1,2-a]pyridine dihydrochloride

  • MDL number MFCD11505517
  • Cas Number : 1171091-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181...183
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39760

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39760
Availability details for
Quantity
  • Availability
  • Add to Cart