ENA183042124 Enamine

4-[(pyrimidin-2-yl)amino]butanoic acid

  • MDL number MFCD08445194
  • Cas Number : 27179-33-5
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39329

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39329
Availability details for
Quantity
  • Availability
  • Add to Cart