ENA183042129 Enamine

N-[(furan-2-yl)methyl]-1-methyl-4-nitro-1H-imidazol-5-amine

  • MDL number MFCD01086090
  • Cas Number : 514200-90-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39337

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39337
Availability details for
Quantity
  • Availability
  • Add to Cart