ENA183042133 Enamine

5-(2-fluorobenzyl)-1,3-thiazol-2-amine

  • MDL number MFCD05863659
  • Cas Number : 876715-72-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97...99
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36692

Safety Information

msds https://www.enaminestore.com/catalog/EN300-36692
Availability details for
Quantity
  • Availability
  • Add to Cart