ENA183042135 Enamine

2-(5-bromo-1-benzofuran-2-yl)ethan-1-ol

  • MDL number MFCD09029646
  • Cas Number : 460748-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39364

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39364
Availability details for
Quantity
  • Availability
  • Add to Cart