ENA183042140 Enamine

4-(5-amino-4H-1,2,4-triazol-3-yl)benzonitrile

  • MDL number MFCD09811672
  • Cas Number : 168893-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-36993

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36993
Availability details for
Quantity
  • Availability
  • Add to Cart