ENA183042141 Enamine

(2-methoxyethyl)(pentan-3-yl)amine

  • MDL number MFCD11505424
  • Cas Number : 1019551-00-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-39370

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39370
Availability details for
Quantity
  • Availability
  • Add to Cart