ENA183042144 Enamine

4-(5-phenyl-1,3,4-oxadiazol-2-yl)butanoic acid

  • MDL number MFCD01244503
  • Cas Number : 322725-59-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39373

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39373
Availability details for
Quantity
  • Availability
  • Add to Cart