ENA183042149 Enamine

2-[2-(propan-2-yl)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD10007075
  • Cas Number : 165315-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39381

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39381
Availability details for
Quantity
  • Availability
  • Add to Cart