ENA183042155 Enamine

2-(2,4-dimethyl-6-oxo-1,6-dihydropyrimidin-5-yl)acetic acid

  • MDL number MFCD08753434
  • Cas Number : 5536-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 240-241
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37022

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37022
Availability details for
Quantity
  • Availability
  • Add to Cart