ENA183042160 Enamine

2-(2-methylpropoxy)propanoic acid

  • MDL number MFCD09929435
  • Cas Number : 53103-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39395

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39395
Availability details for
Quantity
  • Availability
  • Add to Cart