ENA183042161 Enamine

2-{4H,5H,6H,7H-thieno[3,2-c]pyridin-5-yl}aniline

  • MDL number MFCD09045796
  • Cas Number : 926210-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39396

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39396
Availability details for
Quantity
  • Availability
  • Add to Cart