ENA183042164 Enamine

1-{2-[(morpholine-4-sulfonyl)methyl]phenyl}methanamine

  • MDL number MFCD08444208
  • Cas Number : 923213-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39401

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39401
Availability details for
Quantity
  • Availability
  • Add to Cart