ENA183042168 Enamine

2-methanesulfonylpropanoic acid

  • MDL number MFCD09932020
  • Cas Number : 809280-27-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-96
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39405

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39405
Availability details for
Quantity
  • Availability
  • Add to Cart