ENA183042171 Enamine

3-(pyridin-3-ylmethoxy)benzaldehyde oxime

  • MDL number MFCD11505432
  • Cas Number : 1030421-72-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39408

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39408
Availability details for
Quantity
  • Availability
  • Add to Cart