ENA183042173 Enamine

3-acetylbenzene-1-sulfonamide

  • MDL number MFCD06409181
  • Cas Number : 35203-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-138
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-39410

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39410
Availability details for
Quantity
  • Availability
  • Add to Cart