ENA183042175 Enamine

1-(1H-benzimidazol-2-yl)-2-methylpropan-1-amine dihydrochloride

  • MDL number MFCD09753994
  • Cas Number : 25810-65-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37478

Safety Information

msds https://www.enaminestore.com/catalog/EN300-37478
Availability details for
Quantity
  • Availability
  • Add to Cart