ENA183042176 Enamine

2,2,2-trichloro-N-(2,4-difluorophenyl)acetamide

  • MDL number MFCD00751601
  • Cas Number : 349612-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39413

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39413
Availability details for
Quantity
  • Availability
  • Add to Cart