ENA183042183 Enamine

ethyl 4-hydroxy-1-(4-nitrophenyl)-1H-pyrazole-3-carboxylate

  • MDL number MFCD01917145
  • Cas Number : 26502-55-6
  • Molecular Weight : 277.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-39426

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39426
Availability details for
Quantity
  • Availability
  • Add to Cart