ENA183042184 Enamine

3-(pyrimidin-2-ylamino)propanoic acid

  • MDL number MFCD08442108
  • Cas Number : 339195-50-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190...192
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39427

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39427
Availability details for
Quantity
  • Availability
  • Add to Cart