ENA183042185 Enamine

2-(aminosulfonyl)-N,N-dimethylnicotinamide

  • MDL number MFCD03265507
  • Cas Number : 112006-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39428

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-39428
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart