ENA183042194 Enamine

4-(2-morpholin-4-yl-2-oxoethyl)aniline

  • MDL number MFCD09046553
  • Cas Number : 926247-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39440

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39440
Availability details for
Quantity
  • Availability
  • Add to Cart