ENA183042203 Enamine

2-{[4-(pyridin-2-yl)piperazin-1-yl]sulfonyl}ethan-1-amine

  • MDL number MFCD09811110
  • Cas Number : 951909-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39454

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39454
Availability details for
Quantity
  • Availability
  • Add to Cart